EMAIL THIS PAGE TO A FRIEND
Journal of biotechnology
Multifunctional modification of wool using an enzymatic process in aqueous-organic media.
PMID 19428731
Abstract
An enzymatic method using laccases for grafting the water insoluble phenolic compound lauryl gallate on wool fabric was developed. To find the compromise conditions at which the substrate is soluble while the enzyme remains active, the reaction was carried out in an 80/20 (v/v, %) aqueous-ethanol mixture, where the enzyme retains 75-80% of its activity. The enzymatic coating of wool with lauryl gallate provided in a one-step process a multifunctional textile material with antioxidant, antibacterial and water repellent properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48660
Lauryl gallate, antioxidant, ≥99.0% (HPLC)
C19H30O5
53024
Lauryl gallate, analytical standard
C19H30O5