EMAIL THIS PAGE TO A FRIEND
Organic letters
Pd(O2CCF3)2/benzoquinone: a versatile catalyst system for the decarboxylative olefination of arene carboxylic acids.
PMID 19432465
Abstract
A versatile palladium catalyst system was developed to effect the decarboxylative Heck coupling of a variety of arenecarboxylic acids with a wide range of olefins. The key to obtaining the efficient catalyst system is the use of 1-adamatanecarboxylic acid as additive. Alkyl-substituted olefins with coordinating groups were observed to provide significantly improved regioselectivity compared with other alkyl-substituted olefins lacking coordinating groups, and the acetate group of allylic ester was also tolerated in this reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
525049
4-Methoxy-2-methylbenzoic acid, 97%
C9H10O3