EMAIL THIS PAGE TO A FRIEND
Cardiovascular drugs and therapy
Pharmacological approaches to modifying HDL: more basic science to understand HDL metabolism is necessary. Editorial to: "NO-1886 up-regulates Niemann-Pick C1 protein (NPC1) expression through liver X receptor alpha signaling pathway in THP-1 macrophage-derived foam cells" by Xin Ma et al.
PMID 19434486
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1540
NO-1886
C19H20BrN2O4P