EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Concise total synthesis of (+/-)-aspidospermidine via an oxidative Hosomi-Sakurai process.
PMID 19436917
Abstract
A concise total synthesis of (+/-)-aspidospermidine has been achieved in twelve steps from inexpensive 4-ethylphenol, based on an oxidative Hosomi-Sakurai reaction via the concept of "aromatic ring umpolung".