EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis, kinetic studies and pharmacological evaluation of mutual azo prodrugs of 5-aminosalicylic acid for colon-specific drug delivery in inflammatory bowel disease.
PMID 19442419
Abstract
Colon-specific mutual azo prodrugs of 5-aminosalicylic acid with essential amino acids were synthesized for the management of inflammatory bowel disease. The structures were confirmed by elemental and spectral analyses. 85-88% release of 5-aminosalicylic acid was achieved in rat fecal matter with half-lives ranging from 140 to 160 min, following first order kinetics. The prodrugs exhibited comparable ameliorating effect as that of sulfasalazine on trinitrobenzenesulfonic acid-induced experimental colitis in rats with a better safety profile.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1751
D-Phenylalanine, ≥98% (HPLC)
C9H11NO2