EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
A validated SIM GC/MS method for the simultaneous determination of dextromethorphan and its metabolites dextrorphan, 3-methoxymorphinan and 3-hydroxymorphinan in biological matrices and its application to in vitro CYP2D6 and CYP3A4 inhibition study.
PMID 19444798
Abstract
Dextromethorphan is used as a probe drug for assessing CYP2D6 and CYP3A4 activity in vivo and in vitro. A SIM GC/MS method without derivatization for the simultaneous determination of dextromethorphan and its metabolites, dextrorphan, 3-methoxymorphinan and 3-hydroxymorphinan, in human plasma, urine and in vitro incubation matrix was developed and validated. Calibration curves indicated good linearity with a coefficient of variation (r) better than 0.995. The lower limit of quantitation was found to be 10 ng/mL for all analytes in all matrices. Intra-day and inter-day precision for dextromethorphan and its metabolites was better than 9.02 and 9.91%, respectively and accuracy ranged between 91.76 and 106.27%. Recovery for dextromethorphan, its metabolites and internal standard levallorphan was greater than 72.68%. The method has been successfully applied for the in vitro inhibition of metabolism of dextromethorphan by CYP2D6 and CYP3A4 using known inhibitors of CYPs such as quinidine and verapamil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H156
(+)-3-Hydroxymorphinan hydrobromide, neurotrophic to dopaminergic neurons
C16H21NO · HBr