EMAIL THIS PAGE TO A FRIEND
Chemosphere
Synthesis of 2-(4-chlorophenyl)-2-(4-chloro-3-thiophenol)-1,1-dichloroethene (3-SH-DDE) via Newman-Kwart rearrangement - a precursor for synthesis of radiolabeled and unlabeled alkylsulfonyl-DDEs.
PMID 19447466
Abstract
For the first time, a pathway for synthesis of 2-(4-chlorophenyl)-2-(4-chloro-3-thiophenol)-1,1-dichloroethene (3-SH-DDE), is presented. The compound is of particular interest as a precursor for synthesis of alkylsulfonyl-DDE containing different alkyl groups to discover structural activity relationships, and to promote synthesis of radiolabeled methylsulfonyl-DDE. 2-Chloro-5-methylphenol was first methylated and further oxidized to the corresponding benzoic acid. The acid was reduced to the corresponding aldehyde (4-chloro-3-methoxy benzaldehyde) via 4-chloro-3-methoxy-benzene methanol. A lead/aluminium bimetal system was used to carry out the reductive addition of tetrachloromethane to 4-chloro-3-methoxy benzaldehyde to obtain 2,2,2-trichloro-1-(4-chloro-3-methoxyphenyl)ethanol, the desired starting material to synthesize the DDT-analogue (2-(4-chlorophenyl)-2-(4-chloro-3-methoxy-phenyl)-1,1,1-trichloroethane). Elimination of hydrochloric acid and removal of the methyl group led to the 3-OH-DDE. The Newman-Kwart rearrangement was applied to convert 3-OH-DDE to 3-SH-DDE via the dimethylcarbamothioate derivative. 3-SH-DDE is then used as a precursor for the radiolabel synthesis. The overall yield to acquire 3-SH-DDE after 11 steps was 3%. The step with the lowest yield was the DDT-analog synthesis with a yield of 30%. All other step had a yield of >50%. 3-SH-DDE was methylated with (14)C-labeled iodomethane and oxidized by hydrogen peroxide to obtain 3-[(14)C]MeSO(2)-DDE in an overall yield of 30%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
159557
2-Chloro-5-methylphenol, 99%
C7H7ClO