EMAIL THIS PAGE TO A FRIEND
Environmental toxicology
Effects of exposure to oxamyl, carbofuran, dichlorvos, and lindane on acetylcholinesterase activity in the gills of the Pacific oyster Crassostrea gigas.
PMID 19449386
Abstract
Acetylcholinesterase (AChE) activity has been used to test the exposure of mollusk bivalves to pesticides and other pollutants. The Pacific oyster Crassostrea gigas is a species with a worldwide distribution, and it has a high commercial value. The use of this species as a bioindicator in the marine environment, and the use of measurements of AChE activity in tissues of C. gigas require prior evaluation of organisms exposed to several toxic compounds in the laboratory. In our study, the effects of pesticides on AChE activity in the gills and mantle tissues of C. gigas were analyzed by exposing animals to organophosphate (dichlorvos), carbamate (carbofuran and oxamyl), and organochlorine (lindane) pesticides. Adult Pacific oysters were exposed to several concentrations (0.1-200 microM) of dichlorvos, carbofuran, and oxamyl for 96 h, and lindane (1.0 and 2.5 microM) was applied for 12 days. In gill tissues, all pesticides analyzed caused a decrease in AChE activity when compared to the control unexposed group. The mean inhibition concentration (IC(50)) values were determined for dichlorvos, carbofuran, and oxamyl pesticides. Dichlorvos had the highest toxic effect, with an IC(50) of 1.08 microM; lesser effects were caused by oxamyl and carbofuran, with IC(50)s of 1.67 and 3.03 microM, respectively. This study reports the effects of pesticides with several chemical structures and validates measurement of AChE activity in the gill tissues of C. gigas for use in environmental evaluations or food quality tests.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36184
Oxamyl, PESTANAL®, analytical standard
C7H13N3O3S