EMAIL THIS PAGE TO A FRIEND
Anaesthesia
Meta-analysis on the effect of dopexamine on in-hospital mortality.
PMID 19453310
Abstract
The objective of the study was to determine whether dopexamine alters in-hospital mortality. The following databases were searched, Embase (1974-July 2007), Medline (1950-July 2007), CINAHL, PubMed and Cochrane Clinical Register of Controlled Trials (CENTRAL). Two reviewers independently checked the quality of the studies and extracted data. Six randomised controlled trials totalling 935 patients were included. Mortality was not significantly different with dopexamine treatment (relative risk 0.75, 95% confidence interval 0.48-1.18, p = 0.22). In conclusion, dopexamine does not improve in-hospital mortality in patients undergoing major abdominal surgery and in the critically ill.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000612 Dopexamine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
C22H32N2O2 · 2HCl