EMAIL THIS PAGE TO A FRIEND
Glia
Arachidonic acid: a key molecule for astrocyte survival to peroxynitrite.
PMID 19455584
Abstract
Nontoxic concentrations of peroxynitrite (ONOO(-)) nevertheless commit rat astrocytes to mitochondrial permeability transition-dependent toxicity, however prevented by a signaling response driven by arachidonic acid (ARA). The lipid messenger was released upon ONOO(-)-dependent activation of cytosolic phospholipase A(2) and its pharmacological inhibition, or knock-down, was invariably associated with a prompt apoptotic response sensitive to exogenous ARA, but insensitive to other polyunsaturated fatty acids, as eicosapentaenoic or linoleic acid. Interestingly, while microglia also used ARA to cope with ONOO(-), cerebellar granule cells were killed by the same concentrations of ONOO(-) employed in astrocyte/microglia experiments via a mechanism sensitive to inhibition of ARA release. These results collectively support the notion that resistance of glial cells to ONOO(-), a species extensively produced under neuroinflammatory conditions, is largely based on a critical survival signaling triggered by the inflammatory product ARA. In remarkable contrast with these results, the lipid messenger appears to mediate toxicity in neuronal cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A231
Arachidonyl trifluoromethyl ketone, oil, ≥97% (NMR)
C21H31F3O