EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
A feasibility study of differential delivery of levodopa ester and benserazide using site-specific intestinal loops in rats.
PMID 19455628
Abstract
To study the effect of varied intestinal delivery of levodopa ester, levodopa and its butyl ester were administered as bolus or continuous infusions into site-specific, in situ ligated intestinal loops of rats. Benserazide, a carboxylase inhibitor, was not administered, coadministered with ester, or infused into the duodenal loop prior to ester dosing. While the proximal colon minimally absorbed levodopa itself, it substantially absorbed the ester. Coadministration of benserazide and ester at the colon did not increase absorption; however, prior infusion of benserazide into the duodenum enhanced the colonic absorption of ester. Compared to bolus infusion, continuous delivery of the ester resulted in a more sustained levodopa concentration in plasma, and less metabolism into dopamine. The results were repeated for methyl ester, and the relative differences between the results of methyl and butyl esters versus levodopa were similar. The overall results at the duodenum, jejunum, and ileum were also comparable, likewise were those for the proximal, middle, and distal colons. The results of the study are encouraging: a combination of the continuous delivery of levodopa ester with an immediate-release of benserazide optimizes levodopa bioavailability, potentially leading to a much more effective use of levodopa in the treatment of Parkinson's patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1507
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, solid
C10H13NO4 · HCl