EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Diurnal changes in the synthesis of the neurosteroid 7alpha-hydroxypregnenolone stimulating locomotor activity in newts.
PMID 19456382
Abstract
We recently identified 7alpha-hydroxypregnenolone as a novel amphibian neurosteroid stimulating locomotor activity in newts. Because male newts show marked diurnal changes in locomotor activity, we hypothesized that 7alpha-hydroxypregnenolone may be a key factor for the induction of diurnal changes in locomotor activity in male newts. In this study, we found diurnal changes in 7alpha-hydroxypregnenolone synthesis in the brain of male newts, which paralleled locomotor activity. Interestingly, the production of 7alpha-hydroxypregnenolone in the male newt brain increased during the dark phase when locomotor activity of males was high.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H5002
17α-Hydroxypregnenolone
C21H32O3