EMAIL THIS PAGE TO A FRIEND
Cancer letters
Corosolic acid induces apoptosis through mitochondrial pathway and caspase activation in human cervix adenocarcinoma HeLa cells.
PMID 19457606
Abstract
We investigated the response of human cervix adenocarcinoma HeLa cells to Corosolic acid (CRA) treatment. Our results showed that CRA significantly inhibited cell viability in both a dose- and a time-dependent manner. CRA treatment induced S cell-cycle arrest and caused apoptotic death in HeLa cells. We found that CRA increased in Bax/Bcl-2 ratios by up-regulating Bax expression, disrupted mitochondrial membrane potential and triggered the release of cytochrome c from mitochondria into the cytoplasm. Moreover, CRA treatment triggered the activation of caspase-8, -9 and -3 in HeLa cells. All these results indicate that CRA-induced apoptosis is associated with the activation of caspases via a mitochondrial pathway. Taken together, we believe that CRA could have strong potentials for clinical application in treating human cervix adenocarcinoma and improving cancer chemotherapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0549
Corosolic acid from Lagerstroemia speciosa, ≥98% (HPLC)
C30H48O4