EMAIL THIS PAGE TO A FRIEND
Chemical senses
Nearest neural neighbors: moth sex pheromone receptors HR11 and HR13.
PMID 19458025
Abstract
In moth sex pheromone olfaction systems, there is a stereotypical co-compartmentalization of two or sometimes three olfactory receptor neurons (ORNs) within single trichoid sensilla on which pheromone-sensitive odorant receptors (ORs) are differentially expressed. In this issue of Chemical Senses, Krieger et al. show through elegant double and triple in situ hybridization studies that in the moth, Heliothis virescens, a pheromone component-related OR (HR11) is expressed on an ORN that is reliably cocompartmentalized in the same sensillum as another OR (HR13) whose ligand is known to be (Z)-11-hexadecenal, the H. virescens major pheromone component. Although the ligand for HR11 is not yet known, mapping this OR to this particular ORN represents a key advance in piecing together the puzzle of H. virescens sex pheromone olfaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
249084
cis-11-Hexadecenal, 95%
C16H30O