EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Molecular design, device function and surface potential of zwitterionic electron injection layers.
PMID 19459628
Abstract
A series of zwitterionic molecules were synthesized by addition of iodoalkanes to sodium tetrakis(1-imidazolyl)borate (NaBIm(4)). Single substitution at the N3 site leads to compounds designated as C(n)-BIm(4) (n = 1, 6, 12, 16), where the subscript corresponds to the number of carbon atoms in the linear alkyl chain. Metrical parameters in the molecular structure of C(1)-BIm(4) determined by single crystal X-ray diffraction studies confirms the zwitterionic nature and were used to calculate the molecular dipole moment. Compounds C(n)-BIm(4) can be used to improve the electroluminescence efficiencies of polymer light emitting diodes (PLEDs) fabricated by simple solution methods that use poly[2-methoxy-5-(2'-ethylhexyloxy-1,4-phenylenevinylene)] (MEH-PPV) as the emissive semiconducting layer and aluminum as the cathode. This improvement is primarily due to a reduction in the barrier to electron injection and is strongly dependent on the length of the alkyl substituent in C(n)-BIm(4), with C(16)-BIm(4) yielding the best performance. Examination of the surface topography of C(1)-BIm(4) and C(16)-BIm(4) atop MEH-PPV by atomic force microscopy (AFM) shows that the longer alkyl chains assist in adhesion to the surface, however the coverage is poor due to insufficient wetting. These studies also reveal an unexpected planarization of the C(16)-BIm(4) layer adjacent to the electrodes after metal deposition, which is suggested to improve electrical contact. A combination of open circuit voltage measurements, surface reconstruction studies and surface potential measurements indicates that zwitterion insertion leads to the formation of a spontaneously organized dipole layer at the metal/organic interface. The net effect is a shift in the vacuum level and concomitant improvement of electron injection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
279641
Sodium tetrakis(1-imidazolyl)borate, 97%
C12H12BN8Na