EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of amino acid derivatives of quinolone antibiotics.
PMID 19462046
Abstract
Optically pure conjugates of quinolone antibiotics with naturally occurring amino acids are synthesized in 40-98% yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8645
Cinoxacin
C12H10N2O5
O0877
Oxolinic acid, quinolone antibiotic
C13H11NO5
67126
Oxolinic acid, analytical standard
C13H11NO5