EMAIL THIS PAGE TO A FRIEND
Steroids
Oestrogen and progestins differently prevent glutamate toxicity in cortical neurons depending on prior hormonal exposure via the induction of neural nitric oxide synthase.
PMID 19463685
Abstract
Sex steroids are important for brain function and protection. However, growing evidence suggests that these actions might depend on the timing of exposure to steroids. We have studied the effects of steroid administration on the survival of neural cells and we have partially characterized the possible mechanisms. The effect of a 24h pre-treatment with 17beta-estradiol or 17beta-estradiol plus progesterone or medroxyprogesterone acetate on the toxic action of l-glutamate was used to test the experimental hypothesis. Pre-exposure to either steroid combinations turned in enhanced cell survival. Instead, addition of sex steroids together with l-glutamate, in the absence of a pre-exposure had no protective effect. Pre-treatment with the steroid combinations resulted in increased neural NOS expression and activity and blockade of NOS abolished the cytoprotective effects of steroids. These results suggest that NOS induction might be involved in sex steroid-induced neuroprotection. Furthermore, these data supports the hypothesis that prolonged and continued exposure to oestrogen and progesterone, leading to changes in gene expression, is necessary to obtain neuroprotection induced by sex steroids.