EMAIL THIS PAGE TO A FRIEND
FEBS letters
Crystal structure of alpha/beta-galactoside alpha2,3-sialyltransferase from a luminous marine bacterium, Photobacterium phosphoreum.
PMID 19467231
Abstract
Alpha/beta-galactoside alpha2,3-sialyltransferase produced by Photobacterium phosphoreum JT-ISH-467 is a unique enzyme that catalyzes the transfer of N-acetylneuraminic acid residue from cytidine monophosphate N-acetylneuraminic acid to acceptor carbohydrate groups. The enzyme recognizes both mono- and di-saccharides as acceptor substrates, and can transfer Neu5Ac to both alpha-galactoside and beta-galactoside, efficiently. To elucidate the structural basis for the broad acceptor substrate specificity, we determined the crystal structure of the alpha2,3-sialyltransferase in complex with CMP. The overall structure belongs to the glycosyltransferase-B structural group. We could model a reasonable active conformation structure based on the crystal structure. The predicted structure suggested that the broad substrate specificity could be attributed to the wider entrance of the acceptor substrate binding site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8271
Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt, ≥85% (HPLC)
C20H30N4NaO16P