EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Effects of a thiosemicarbazide camphene derivative on Trichophyton mentagrophytes.
PMID 19471200
Abstract
Thiosemicarbazides are compounds known for their biological activity, particularly their antimicrobial properties, which include activity against fungi. The difficulty of treating fungal diseases induced us to assess the antifungal properties of some novel thiosemicarbazide compounds. We selected the natural products limonene and camphene as sources for the preparation of these new thiosemicarbazide derivatives. The compound N(4)-[2,2-dimethyl-3-methylnorbornane]-thiosemicarbazide (TIO C) showed an antifungal effect on Trichophyton mentagrophytes, with values of MIC = 55 mmol L(-1) and MFC = 110 micromol L(-1). Scanning-electron microscopy showed a decrease in mycelium development and morphological alterations of T. mentagrophytes cultured on nail fragments and treated with TIO C. In an attempt to discover its mode of action, we noted that ergosterol is apparently not a target of TIO C activity. An effect of TIO C on T. mentagrophytes cell walls and dividing cross-walls was shown by observed impairment of the fluorescence of tissues stained with calcofluor white, a specific marker for fungal chitin, suggesting that the compound can affect and damage the cell-wall structure or may interfere with its formation, during cell division, growth, and morphogenesis. This approach to the synthesis of new derivatives might provide interesting compounds with greater biological activity in pharmacological research.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442505
Camphene, analytical standard
C10H16
456055
Camphene, 95%
C10H16