EMAIL THIS PAGE TO A FRIEND
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry
DNA interactions and photocatalytic strand cleavage by artificial nucleases based on water-soluble gold(III) porphyrins.
PMID 19471974
Abstract
The novel gold porphyrin complex (5,10,15-tris(N-methylpyridinium-4-yl)-20-(1-pyrenyl)-porphyrinato)gold(III) chloride, [Au(III)(TMPy3Pyr1P)]Cl4, was prepared and characterized by optical spectroscopy, high-resolution nuclear magnetic resonance (NMR), and electrospray mass spectrometry. This cationic multichromophore compound exhibits excellent water solubility and does not form aggregates under physiological conditions. Binding interactions of this complex and related model compounds with nucleic acid substrates have been studied and characterized by NMR and circular dichroism spectroscopy. The photoreactivity of [Au(III)(TMPy3Pyr1P)]Cl4 was investigated under anaerobic and aerobic conditions in the presence of an excess of purine nucleoside, guanosine, and plasmid DNA. Photocatalytic oxidative degradation of guanosine and the change from supercoiled to circular plasmid DNA upon monochromatic irradiation and polychromatic blue-light exposure with a maximum at 420 nm was explored. The potential of the novel water-soluble cationic metallointercalator complex [Au(III)(TMPy3Pyr1P)]Cl4 to serve as a catalytic photonuclease for the cleavage of DNA has been demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852228
2′-Deoxyguanosine 5′-monophosphate disodium salt hydrate, ≥98%
C10H12N5Na2O7P · xH2O