EMAIL THIS PAGE TO A FRIEND
Organic letters
X-ray structural characterization of charge delocalization onto the three equivalent benzenoid rings in hexamethoxytriptycene cation radical.
PMID 19473043
Abstract
Definitive X-ray crystallographic evidence is obtained for a single hole (or a polaron) to be uniformly distributed on the three equivalent 1,2-dimethoxybenzenoid (or veratrole) rings in the hexamethoxytriptycene cation radical. This conclusion is further supported by electrochemical analysis and by the observation of an intense near-IR transition in its electronic spectrum, as well as by comparison of the spectral and electrochemical characteristics with the model compounds containing one and two dimethoxybenzene rings.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
140155
1,2-Dimethoxybenzene, ReagentPlus®, 99%
C8H10O2
W379905
1,2-Dimethoxybenzene, ≥99%, FG
C8H10O2