EMAIL THIS PAGE TO A FRIEND
Journal of human genetics
A two-stage case-control association study of PADI2 with schizophrenia.
PMID 19478818
Abstract
Peptidylarginine deiminases (PADIs), five isoforms of which have been identified, catalyze the conversion of arginine residues to citrulline residues in proteins. Recent studies have revealed that abnormal activation of PADI2, the gene for which is expressed throughout the nervous system, is likely to be related to the pathogenesis of neuropsychiatric diseases with neurodegenerative processes, such as Alzheimer's disease and multiple sclerosis. Such a progressive neurodegenerative process could be involved in the etiology and/or course of schizophrenia, and PADI2 may be a candidate gene for schizophrenia. To assess whether PADI2 has a role in vulnerability to schizophrenia, we conducted a two-stage case-control association study in Japanese individuals. In a screening population of 534 patients and 559 control individuals, we examined eight single-nucleotide polymorphisms (SNPs) including four haplotype tag SNPs and four coding SNPs in PADI2. There was a potential association of a synonymous SNP in exon 7 with schizophrenia. However, we could not replicate this association in a confirmatory population of 2126 patients and 2228 control individuals. The results of this study suggest that PADI2 does not contribute to genetic susceptibility to schizophrenia.
Related Materials