EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Facile synthesis and in vitro properties of 1-alkyl- and 1-alkyl-N-propargyl-1,2,3,4-tetrahydroisoquinoline derivatives on PC12 cells.
PMID 19481842
Abstract
The synthesis of several 1-alkyl-1,2,3,4-tetrahydroisoquinolines, which may play an important role in Parkinson's disease, has been achieved by modified Pictet-Spengler cyclization of the formyliminium ion. The direct cytotoxicity and preventative effects towards MPP(+)-mediated death of PC12 cells were estimated. The cytotoxicities of 1-alkyl-TIQs were apoptotic and depended on their lipophilic properties. Conversely, introducing the N-propargyl substituent reduced cytotoxicity. 1-Methyl-, 1-methyl-N-propargyl- and 1-cyclopropyl-TIQ partially inhibited MPP(+)-induced cell death, whereas relatively large alkyl substituents at the first position did not enhance the viability of PC12 cells. In summary, our findings suggest a crucial role for the N-propargyl functional group for the effective reduction of cytotoxicity, and show the importance of size and lipophilicity of substituents at the 1-position of 1-alkyl-TIQs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T13005
1,2,3,4-Tetrahydroisoquinoline, 95%
C9H11N