EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Identification of a novel self-sufficient styrene monooxygenase from Rhodococcus opacus 1CP.
PMID 19482928
Abstract
Sequence analysis of a 9-kb genomic fragment of the actinobacterium Rhodococcus opacus 1CP led to identification of an open reading frame encoding a novel fusion protein, StyA2B, with a putative function in styrene metabolism via styrene oxide and phenylacetic acid. Gene cluster analysis indicated that the highly related fusion proteins of Nocardia farcinica IFM10152 and Arthrobacter aurescens TC1 are involved in a similar physiological process. Whereas 413 amino acids of the N terminus of StyA2B are highly similar to those of the oxygenases of two-component styrene monooxygenases (SMOs) from pseudomonads, the residual 160 amino acids of the C terminus show significant homology to the flavin reductases of these systems. Cloning and functional expression of His(10)-StyA2B revealed for the first time that the fusion protein does in fact catalyze two separate reactions. Strictly NADH-dependent reduction of flavins and highly enantioselective oxygenation of styrene to (S)-styrene oxide were shown. Inhibition studies and photometric analysis of recombinant StyA2B indicated the absence of tightly bound heme and flavin cofactors in this self-sufficient monooxygenase. StyA2B oxygenates a spectrum of aromatic compounds similar to those of two-component SMOs. However, the specific activities of the flavin-reducing and styrene-oxidizing functions of StyA2B are one to two orders of magnitude lower than those of StyA/StyB from Pseudomonas sp. strain VLB120.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05470
Bovine serum albumin, lyophilized powder, crystallized, ≥98.0% (GE)
A4503
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
A7888
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
A9647
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
A3294
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
A6003
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
A3059
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
A0281
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
A7638
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
A8022
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
A3912
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
A3803
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
B4287
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
A8806
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
A4378
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
A7511
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized
A4919
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
A2058
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
A3311
Bovine Serum Albumin, powder, BioXtra
A1933
Bovine Serum Albumin, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
A4161
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
A9430
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
A2934
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
B6917
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
A3156
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
A9306
Bovine Serum Albumin, lyophilized powder, low endotoxin, ≥95% (agarose gel electrophoresis)
A9543
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
A3675
Bovine Serum Albumin, lyophilized powder, low endotoxin, ≥98% (agarose gel electrophoresis)
A9056
Bovine Serum Albumin, lyophilized powder, Microbiologically tested, ≥96% (agarose gel electrophoresis)
A9085
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)