EMAIL THIS PAGE TO A FRIEND
The American Journal of Psychiatry
Differential expression of metabotropic glutamate receptors 2 and 3 in schizophrenia: a mechanism for antipsychotic drug action?
PMID 19487395