EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Comparative analysis of odorous volatile organic compounds between direct injection and solid-phase microextraction: development and validation of a gas chromatography-mass spectrometry-based methodology.
PMID 19493534
Abstract
In this study, the feasibility of GC-MS was evaluated for the quantification of odorous volatile organic compounds (VOCs) in environmental samples. These included methyl ethyl ketone, isobutyl alcohol, methyl isobutyl ketone, and butyl acetate plus benzene, toluene, and xylene (BTX). For this purpose, the gaseous standard for these VOCs were analyzed by GC-MS with the aid of both direct injection (DI) into the GC injector and solid-phase microextraction (SPME). The liquid phase standard prepared independently was tested additionally by the DI method as a reference to gaseous calibration. The detection limit (DL) values, when tested for basic quality assurance in this study, showed large differences between DI (0.002-0.007 ng) and SPME method (1.03-1.81 ng) in terms of absolute mass. The DL values, when expressed in terms of concentration (v/v), showed considerable improvement in SPME (below 0.40 nmol mol(-1)) relative to the DI method (approximately 6-15 nmol mol(-1)). The reliability of the GC-MS method was further validated through an analysis of real environmental samples collected from an industrial area.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57318 SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, for use with manual holder, needle size 24 ga
57344-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, needle size 23 ga, for use with manual holder
57319 SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, needle size 24 ga, for use with autosampler
57343-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm, needle size 23 ga, for use with autosampler
57334-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 24 ga, for use with manual holder, StableFlex fiber
57335-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 24 ga, StableFlex, for use with autosampler
57295-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 23 ga, StableFlex, for use with autosampler
57906-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, for use with autosampler, needle size 23 ga, metal alloy fiber
FFA57295-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, needle size 23 ga, Stableflex, for use with multi fiber exchanger
FFA57335-U SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm, for use with multi fiber exchanger, needle size 24 ga, StableFlex
504831 SPME Portable Field Sampler, coating Carboxen/Polydimethylsiloxane (CAR/PDMS)