EMAIL THIS PAGE TO A FRIEND
Current drug discovery technologies
Development of a yeast bio-assay to screen anti-mitochondrial drugs.
PMID 19496752
Abstract
Previous studies show that acetylsalicylic acid (aspirin) at low concentrations affects yeast sexual structure development in a similar fashion than oxygen depletion. This is ascribed to its anti-mitochondrial action. In this study, we report the same for other anti-inflammatory (i.e. ibuprofen, indomethacin, salicylic acid, benzoic acid) as well as anticancer (Lonidamine) drugs, also known for inhibiting mitochondrial activity in mammalian cells. This is shown by a unique yeast bio-assay, with the mitochondrion-dependent sexual structure, riboflavin production, and hyphal morphology of the yeast Eremothecium ashbyi serving as indicators. These drugs affect this yeast in a similar way as found under oxygen limitation conditions by inhibiting sexual structure development (most sensitive), riboflavin production, and yielding characteristically wrinkled and granular hyphae, presenting a unique "anoxic" morphological pattern for this yeast. Only drugs associated with anti-mitochondrial activity presented such a pattern. This bio-assay may find application in the screening for novel drugs from various sources with anti-mitochondrial actions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L4900
Lonidamine, mitochondrial hexokinase inhibitor
C15H10Cl2N2O2