EMAIL THIS PAGE TO A FRIEND
The American journal of emergency medicine
Rodenticide poisoning as a cause of quadriparesis: a rare entity.
PMID 19497472
Abstract
Acute flaccid paralysis with its wide repertoire of causes presents a challenge to the clinician. We present a case of barium carbonate poisoning, a rare yet easily treatable cause of acute quadriparesis. Barium acts by redistribution of potassium from the extra- to intravascular compartment causing hypokalemia. Early recognition and prompt potassium supplementation under cardiac monitoring yield gratifying results.