EMAIL THIS PAGE TO A FRIEND
Stroke
Angiotensin AT2 receptor-mediated neuroprotection and nitric oxide-bioavailability in stroke.
PMID 19498178
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C160
CGP-42112A, ≥95%, synthetic, solid
C52H69N13O11