EMAIL THIS PAGE TO A FRIEND
Biomaterials
Nitric oxide-releasing S-nitrosothiol-modified xerogels.
PMID 19501904
Abstract
The synthesis, material characterization, and in vitro biocompatibility of S-nitrosothiol (RSNO)-modified xerogels are described. Thiol-functionalized xerogel films were formed by hydrolysis and co-condensation of 3-mercaptopropyltrimethoxysilane (MPTMS) and methyltrimethoxysilane (MTMOS) sol-gel precursors at varying concentrations. Subsequent thiol nitrosation via acidified nitrite produced RSNO-modified xerogels capable of generating nitric oxide (NO) for up to 2 weeks under physiological conditions. Xerogels also exhibited NO generation upon irradiation with broad-spectrum light or exposure to copper, with NO fluxes proportional to wattage and concentration, respectively. Xerogels were capable of storing up to approximately 1.31 micromol NO mg(-1), and displayed negligible fragmentation over a 2-week period. Platelet and bacterial adhesion to nitrosated films was reduced compared to non-nitrosated controls, confirming the antithrombotic and antibacterial properties of the NO-releasing materials. Fibroblast cell viability was maintained on the xerogel surfaces illustrating the promise of RSNO-modified xerogels as biomedical device coatings.