EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Structure-based rational design of a phosphotriesterase.
PMID 19502439
Abstract
In silico substrate docking of both stereoisomers of the pesticide chlorfenvinphos (CVP) in the phosphotriesterase from Agrobacterium radiobacter identified two residues (F131 and W132) that prevent productive substrate binding and cause stereospecificity. A variant (W131H/F132A) was designed that exhibited ca. 480-fold and 8-fold increases in the rate of Z-CVP and E-CVP hydrolysis, respectively, eliminating stereospecificity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36551
Chlorfenvinphos, PESTANAL®, analytical standard
C12H14Cl3O4P