EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Self-assembly of porphyrin-azulene-porphyrin and porphyrin-azulene conjugates.
PMID 19503927
Abstract
In this paper we report the synthesis and self-assembling behavior of new porphyrin-azulene-porphyrin and porphyrin-azulene conjugates. The porphyrin-azulene-porphyrin conjugate gelates a number of organic solvents, while the porphyrin-azulene conjugates form vesicles in a chloroform-methanol binary mixture. The structures of the organogels and vesicles have been characterized by SEM and AFM. Two porphyrin-naphthalene-porphyrin and porphyrin-naphthalene conjugates were also prepared. A comparison of their properties with those of the azulene analogues reveals that the intermolecular dipole-dipole interaction of the azulene units plays an important role in promoting the self-assembly of the porphyrin-azulene-porphyrin and porphyrin-azulene conjugates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37879
Azulene, analytical standard
C10H8
A97203
Azulene, 99%
C10H8