EMAIL THIS PAGE TO A FRIEND
International journal of antimicrobial agents
In vitro and ex vivo activity of peptide deformylase inhibitors against Mycobacterium tuberculosis H37Rv.
PMID 19505802
Abstract
Bacterial peptide deformylase (PDF) catalyses removal of the N-terminal formyl group of proteins and is essential for protein maturation, growth and survival of bacteria. Thus, PDF appears to be a good antimycobacterial drug target. In the present study, various well-known PDF inhibitors, such as BB-3497, actinonin, 1,10-phenanthroline, hydroxylamine hydrochloride and galardin, were selected to evaluate their inhibitory activity against Mycobacterium tuberculosis. All compounds were found to be active against M. tuberculosis, with MIC(90) values (lowest drug concentration at which 90% of growth was inhibited on the basis of CFU enumeration) ranging from 0.2 mg/L to 74 mg/L. BB-3497 and 1,10-phenanthroline exhibited potent in vitro antimycobacterial activity, and also showed synergism with isoniazid and rifampicin. All compounds showed a bacteriostatic mode of inhibition. Under ex vivo conditions and short-course chemotherapy, BB-3497 and actinonin were found to be significantly active, with BB-3497 exhibiting comparable efficacy to that of isoniazid. Collectively, promising activities of PDF inhibitors such as BB-3497 and actinonin suggest their potential use against M. tuberculosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6671
Actinonin
C19H35N3O5