EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Fe(II) phthalocyanine catalyzed oxidation of dGMP by molecular oxygen.
PMID 19505823
Abstract
We show that iron(II)-phthalocyanines are able to catalyze guanosine oxidation by molecular oxygen in the presence of reducing agents such as ascorbic acid and 2-mercaptoethanol. The products of 5'-monophosphate-2'-deoxyguanosine (dGMP) oxidation were directly analyzed using the HPLC-ESI/MS method. The main oxidation products were 5'-phospho-2'-deoxy-8-oxo-7,8-dihydroguanine and the 1,N2-glyoxal adduct of the 5'-monophosphate-2'-deoxyguanosine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852228
2′-Deoxyguanosine 5′-monophosphate disodium salt hydrate, ≥98%
C10H12N5Na2O7P · xH2O