EMAIL THIS PAGE TO A FRIEND
Mutagenesis
Plumbagin induces cell death through a copper-redox cycle mechanism in human cancer cells.
PMID 19505895
Abstract
Plumbagin, a naphthoquinone derived from the medicinal plant Plumbago zeylanica has been shown to exert anticancer and anti-proliferative activities in cells in culture as well as animal tumor models. In our previous paper, we have reported the cytotoxic action of plumbagin in plasmid pBR322 DNA as well as human peripheral blood lymphocytes through a redox mechanism involving copper. Copper has been shown to be capable of mediating the action of several plant-derived compounds through production of reactive oxygen species (ROS). The objective of the present study was to determine whether plumbagin induces apoptosis in human cancer cells through the same mechanism which we proposed earlier. Using 3-(4,5-dimethylthiazol-2-yl)-5-(3-carboxymethoxyphenyl)-2-(4-sulfophenyl)-2H-tetrazolium, inner salt assay, 3-(4,5-B-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay for cell growth inhibition, histone/DNA ELISA, homogeneous caspase-3/7 assay for apoptosis as well as alkaline comet assay for DNA single-strand breaks detection in this report, we confirm that plumbagin causes effective cell growth inhibition, induces apoptosis and generates single-strand breaks in cancer cells. Incubation of cancer cells with scavengers of ROS and neocuproine inhibited the cytotoxic action of plumbagin proving that generation of ROS and Cu(I) are the critical mediators in plumbagin-induced cell growth inhibition. This study is the first to investigate the copper-mediated anticancer mechanism of plumbagin in human cancer cells and these properties of plumbagin could be further explored for the development of anticancer agents with higher therapeutic indices, especially for skin cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1501
Neocuproine, ≥98%
C14H12N2