EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Studying the photoprotective activity of a new class of heteroaromatic antioxidants.
PMID 19513419
Abstract
Photoprotective activity of heteroaromatic compounds (derivatives of 3-hydroxypyridine, amino-6-hydroxybenzothiazole, and 5-hydroxybenzimidazole) was studied in the system of UV-induced cardiolipin peroxidation. Although all three compounds had the antioxidant effect during free radical oxidation of luminol, only derivatives of amino-6-hydroxybenzothiazole and 5-hydroxybenzimidazole inhibited the process of UV-induced lipid peroxidation. The 3-hydroxypyridine derivative did not inhibit UV-induced cardiolipin peroxidation, which was probably related to degradation of this compound under the influence of UV light and formation of degradation products that cannot inhibit free radical processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H57009
3-Hydroxypyridine, 98%
C5H5NO