EMAIL THIS PAGE TO A FRIEND
Cutaneous and ocular toxicology
Inhalational and dermal injury due to explosion of calcium hypochlorite.
PMID 19514924
Abstract
Calcium hypochlorite is a yellow-white powder widely used as a disinfectant in swimming pools. It releases chlorine gas when added to water and can cause respiratory effects. Dermal and eye injury can occur because of the caustic nature of chlorine. We report a case of chlorine toxicity and burns on a man's face due to the explosion of calcium hypochlorite while he was mixing it into the water.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
211389
Calcium hypochlorite, technical grade
CaCl2O2