EMAIL THIS PAGE TO A FRIEND
Bioorganic & Medicinal Chemistry
Molecular modifications on carboxylic acid derivatives as potent histone deacetylase inhibitors: Activity and docking studies.
PMID 19520580
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
205664
1′-Hydroxy-2′-acetonaphthone, 99%
C12H10O2
126209
2,3-Dihydroxybenzoic acid, 99%
C7H6O4
W392103
2,4-Hexadienoic acid, ≥99%, FCC
C6H8O2
102601
3,4-Dihydroxyhydrocinnamic acid, 98%
C9H10O4
252565
8-Hydroxyquinoline, ACS reagent, 99%
C9H7NO
H6878
8-Hydroxyquinoline, crystalline
C9H7NO
36524
8-Quinolinol, PESTANAL®, analytical standard
C9H7NO
55070
8-Quinolinol, puriss. p.a., ACS reagent, for the detection and determination of Al, Mg and others, ≥99.0% (NT)
C9H7NO
32502
8-Quinolinol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (perchloric acid titration)
C9H7NO
10680
Anthranilic acid, puriss. p.a., ≥99.5% (T)
C7H7NO2
A89855
Anthranilic acid, reagent grade, ≥98%
C7H7NO2
B5194
Benzilic acid, ≥99%
C14H12O3
T206
D-(−)-Tartaric acid, ReagentPlus®, 99%
C4H6O6
483796
D-(−)-Tartaric acid, 99%
C4H6O6
95320
D-(−)-Tartaric acid, puriss., unnatural form, ≥99.0% (T)
C4H6O6
M2625
DL-Mandelic acid
C8H8O3
03918
DL-Tartaric acid, anhydrous, for ion chromatography, ≥99.5% (T)
C4H6O6
T400
DL-Tartaric acid, ReagentPlus®, 99%
C4H6O6
W304401
DL-Tartaric acid, ≥99%
C4H6O6
50433
DL-Tartaric acid concentrate, 0.1 M HOOC(CHOH)2COOH (0.2N), eluent concentrate for IC
C4H6O6
I5109
Indole-2-carboxylic acid, 98%
C9H7NO2
I1754
Isobutyric acid, 99%
C4H8O2
58360
Isobutyric acid, puriss. p.a., ≥99.5%
C4H8O2
W222208
Isobutyric acid, ≥99%, FCC, FG
C4H8O2
W222216
Isobutyric acid, natural, ≥99%, FCC, FG
C4H8O2
129542
Isovaleric acid, 99%
C5H10O2
W310204
Isovaleric acid, ≥99%, FCC, FG
C5H10O2
W310212
Isovaleric acid, natural, ≥98%, FG
C5H10O2
K3125
Kojic acid
C6H6O4
M2101
Mandelic acid, 99%
C8H8O3