EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
The first bioreversible prodrug of metformin with improved lipophilicity and enhanced intestinal absorption.
PMID 19522462
Abstract
Metformin is a potent antidiabetic agent and currently used as a first-line treatment for patients with type 2 diabetes. Unfortunately, the moderate absorption and uncomfortable gastrointestinal adverse effects associated with metformin therapy impair its use. In this study, two novel prodrugs of a biguanidine functionality containing antidiabetic agent, metformin, were designed, synthesized, and evaluated in vitro and in vivo to accomplish improved lipophilicity and, consequently, enhanced oral absorption of this highly water-soluble drug. These results represent that the more lipophilic prodrug 2a biotransformed quantitatively to metformin mainly after absorption. The enhanced oral absorption consequently promoted the bioavailability of metformin from 43% to 65% in rats. Thus, this novel prodrug may offer a solution to reduce the required daily doses of metformin, which may decrease the uncomfortable adverse effects associated with metformin therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8876
Sulfamerazine, ReagentPlus®, ≥99.0%
C11H12N4O2S
46826
Sulfamerazine, VETRANAL, analytical standard
C11H12N4O2S