EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis of 9-beta-d-arabinofuranosylguanine by combined use of two whole cell biocatalysts.
PMID 19523826
Abstract
Unlike the preparation of other purine nucleosides, transglycosylation from a pyrimidine nucleoside and guanine is difficult because of the low solubility of this base. Thus, another strategy, based on the coupled action of two whole cell biocatalyzed reactions, transglycosylation and deamination, was used. Enterobacter gergoviae and Arthrobacter oxydans were employed to synthesize 9-beta-d-arabinofuranosylguanine (AraG), an efficient anti leukemic drug.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4233
Ara-G hydrate, ≥98% (HPLC), solid
C10H13N5O5 · xH2O