EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and antimultidrug resistance evaluation of icariin and its derivatives.
PMID 19523827
Abstract
A series of icariin derivatives were synthesized. Their multidrug resistance (MDR) reversal activities were evaluated by MTT assay and the results indicated that the derivatives were the potent modulators of MDR. It was showed that the derivatives significantly increased the intracellular accumulation of ADR in MCF-7/ADR cells compared with drug sensitive MCF-7 cells. The results of bi-directional assay and reverse transcription polymerase chain reaction (RT-PCR) assay showed that the derivatives had high inhibitory activity against P-gp efflux function and significantly down-regulated on the expression of P-gp.
Related Materials