EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Simultaneous determination of 2-furfural and 5-methyl-2-furfural using corona discharge ion mobility spectrometry.
PMID 19531891
Abstract
A novel technique, corona discharge ion mobility spectrometry (CD-IMS), was developed for the qualitative and quantitative determination of 2-furfural (F) and 5-methyl-2-furfural (MF) in aqueous solutions. The limits of detection (LODs) were 5.3 x 10(-3) microg/mL for F and 6.7 x 10(-3) microg/mL for MF. The linear dynamic ranges of 1.16 x 10(-2) to 1.04 microg/mL and 2.20 x 10(-2) to 1.10 microg/mL were obtained for F and MF, respectively. The relative standard deviation was below 12% for both compounds. In addition to analysis of the individual compound, simultaneous determination of F and MF was also investigated. It was realized that F imposes a matrix effect on the MF signal and vice versa. The standard addition method was used to deal with the matrix effect. The recovery of the compounds in the synthetic samples validates the capability of the method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W270202
5-Methylfurfural, ≥98%, FG
C6H6O2
137316
5-Methylfurfural, ReagentPlus®, 99%
C6H6O2