EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Glycosidase inhibition by ring-modified castanospermine analogues: tackling enzyme selectivity by inhibitor tailoring.
PMID 19532990
Abstract
Synthesis of a panel of iso(thio)urea-type ring-modified castanospermine analogues bearing a freely mutarotating pseudoanomeric hydroxyl group results in tight-binding beta-glucosidase inhibitors with unusual binding signatures; the presence of an N-octyl substituent imparts a remarkable anomeric selectivity, promoting strong binding of the appropriate beta-anomer by the beta-glucosidase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
532673
(1S,6S,7R,8R,8aR)-1,6,7,8-Tetrahydroxyindolizidine, 98%
C8H15NO4