EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Cytotoxic triterpenoids from the stems of Microtropis japonica.
PMID 19534471
Abstract
Bioassay-guided fractionation of a methanol extract obtained from stems of Microtropis japonica led to the isolation of six new ursane-type triterpenoids (1-6) and a new 2,3-seco-oleanane-type triterpenoid (7), together with seven known compounds. The structures of the new compounds were elucidated using spectroscopic data analysis. Among the known compounds isolated, the main component, 8 (ursolic acid), was active for HL60 cells, and its effects on histone hyperacetylation and the inhibition of histone deacetylase (HDAC) activity were investigated.