EMAIL THIS PAGE TO A FRIEND
Biomedica biochimica acta
Lipoxygenase-inhibitory action of antiviral polymeric oxidation products of polyphenols.
PMID 1953697
Abstract
Various polymeric oxidation products of polyphenols strongly inhibited the purified lipoxygenase of rabbit reticulocytes, whereas the prostaglandin H synthase of sheep vesicular gland was only weakly inhibited. The oxidized polymeric preparations of caffeic acid, 2,5-dihydroxytoluene and 3,4-dihydroxytoluene were the most potent lipoxygenase inhibitors. Since such preparations are also known to inhibit herpes simplex virus in an early stage of the virus-cell interaction it is proposed that lipoxygenase metabolites are involved in virus-induced changes in the cell metabolism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
112968
Methylhydroquinone, 99%
C7H8O2
89600
Methylhydroquinone, purum, ≥98.0% (HPLC)
C7H8O2