EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Oncostatin M stimulates urokinase-type plasminogen activator activity in human synovial fibroblasts.
PMID 1953736
Abstract
Cytokine regulation of synovial cell function has been considered to be involved in the pathogenesis of rheumatoid arthritis. Synoviocyte urokinase-type plasminogen activator (u-PA) expression may be relevant to the tissue remodelling, as well as to the cell migration and transformation occurring in rheumatoid joints. We report here that purified recombinant human oncostatin M (greater than or equal to approximately 0.2 U/ml = 1 pM) stimulated within six hr the u-PA activity of non-rheumatoid synovial fibroblast-like cells and raised their u-PA mRNA levels. Oncostatin M could augment PGE2 production and DNA synthesis in these cells; however, the increase in PGE2 was small compared with that caused by IL-1. Since oncostatin M is produced by immune cells, it may have a role in immune and inflammatory reactions by interacting with fibroblast populations, such as synoviocytes, in the manner described.