EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Oxazoline-oxazinone oxidative rearrangement. divergent syntheses of (2S,3S)-4,4,4-trifluorovaline and (2S,4S)-5,5,5-Trifluoroleucine.
PMID 19537688
Abstract
Stereoselective syntheses of the valuable fluorinated amino acids (2S,3S)-4,4,4-trifluorovaline and (2S,4S)-5,5,5-trifluoroleucine have been achieved starting from 4,4,4-trifluoro-3-methylbutanoic acid by using a conceptually simple transformation: conversion to a chiral oxazoline, SeO2-promoted oxidative rearrangement to the dihydro-2H-oxazinone, and face-selective hydrogenation of the C=N bond, followed by hydrogenolysis-hydrolysis. The transformation is limited by the tendency of the intermediate beta-trifluoromethyldihydrooxazinone to undergo imine-enamine isomerization. Both amino acids were obtained as configurationally pure hydrochloride salts identical in all respects with those in literature reports.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91917
5,5,5-Trifluoro-DL-leucine, ≥98.0% (sum of isomers, HPLC)
C6H10F3NO2