EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Rate of loss of simazine, terbuthylazine, isoproturon, and methabenzthiazuron during soil solarization.
PMID 19537795
Abstract
This paper reports the use of solar heating by polyethylene mulching for decontamination of a silty clay-loam soil polluted with herbicides. Soil solarization, a natural and hydrothermal method commonly used for disinfesting soils, was tested during the summer season on a Hipercalcic Calcisol located in Murcia (southeast Spain) for dissipation of s-triazine (simazine and terbuthylazine) and phenylurea (isoproturon and methabenzthiazuron) herbicides using low-density (LD) and high-density (HD) polyethylene (PE) film as a cover. A well-established influence of the film was observed on the dissipation of all herbicides from the soil, although the density (0.92-0.95 g/cm(3)) of the film used (LDPE and HDPE) was not significant in terms of the rate of loss. In all cases, a quick depletion during the first 2 weeks was observed, mainly for terbuthylazine. The first-order model satisfactorily explained the dissipation process, but the Hoerl and biexponential equations were more appropriate, mainly for simazine, isoproturon, and methabenzthiazuron. In all cases, herbicides disappeared at faster rates in solarized soils (DT(50) = 4-29 days) than in nonmulched soils (DT(50) = 11-35 days), especially for terbuthylazine and isoproturon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36156
Methabenzthiazuron, PESTANAL®, analytical standard
C10H11N3OS