EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Estimation of peptide concentration by a modified bicinchoninic acid assay.
PMID 19539599
Abstract
Although biuret based protein assays are theoretically applicable to peptide measurement, there is a high level of interpeptide variation, determined largely by peptide hydrophobicity. This variation in peptide reactivity can be significantly reduced by heat-denaturation of peptides at 95 degrees C for 5 min in the presence of 0.1 M NaOH containing 1% (w/v) SDS, prior to incubation for 30 min at 37 degrees C in BCA standard working reagent. This modification to the standard bicinchoninic acid (BCA) assay protocol allows for an accurate, rapid, and economical estimation of the peptide concentration within an unknown sample.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14335
2,2′-Biquinoline-4,4′-dicarboxylic acid, ≥90% (TLC)
C20H12N2O4