EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Allosteric interaction of the anticholinergic drug [N-(4-phenyl)-phenacyl-l-hyoscyamine] (Phenthonium) with nicotinic receptors of post-ganglionic sympathetic neurons of the rat vas deferens.
PMID 19540221
Abstract
Phenthonium (Phen), a quaternary analog of hyoscyamine, is a blocker of muscarinic activity and an allosteric blocker of alpha(1)2betagammaepsilon nicotinic receptors. Specifically, Phenthonium increases the spontaneous release of acetylcholine at the motor endplate without depolarizing the muscle or inhibiting cholinesterase activity. This paper compares Phenthonium's effects on sympathetic transmission and on ganglionic nicotinic receptor activation. Neurotransmitter release and twitch of the rat vas deferens were induced either by electrical stimulation or by 1,1-dimethyl-4-phenylpiperazine (DMPP) activation of nicotinic receptors. Contractions independent of transmitter release were induced by noradrenaline and adenosine 5'-triphosphate (ATP). Phenthonium inhibited transmitter release and depressed twitch without changing the responsiveness to noradrenaline or ATP. Twitch depression did not occur after K(+)-channel blockade with 4-aminopyridine (4-AP) or charybdotoxin. DMPP had a similar effect, but high concentrations induced contraction of non-stimulated organs. Incubation of Phenthonium inhibited further DMPP twitch depression and non-competitively depressed the contractile responses elicited by DMPP. Furthermore, mecamylamine, but neither methyllycaconitine nor atropine, blocked the contraction elicited by DMPP. Phenthonium and DMPP are K(+)-channel openers that primarily inhibit sympathetic transmission. Contraction induced by DMPP was probably mediated by neuronal nicotinic receptor other than the alpha7 subtype. The blockade of DMPP contractile response was unrelated to Phenthonium's antimuscarinic or K(+)-channel opening activities. Since Phenthonium's quaternary chemical structure limits its membrane diffusion, the non-competitive inhibition of DMPP excitatory responses should be linked to allosteric interaction with neuronal nicotinic receptors that putatively qualify Phenthonium as a novel modulator of cholinergic synapses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5891
1,1-Dimethyl-4-phenylpiperazinium iodide, ≥98% (TLC or titration), powder
C12H19IN2